BRAND® Sealing Mats for Deep-Well Plates
Provides effective plug sealing for each well
  • Reduces evaporation and contamination
Use these flexible deep-well sealing mats to reduce maximum well volume. The mats are made of silicone, polypropylene, or modified polyethylene. Match mat to proper plate for best seal.
$1,059.37CAD / Pkg of 50
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Number of Wells Well Volume (mL)
Availability Pricing
BRAND 701357 Sealing Mat for Deep Well Plate, 384-Well, Silicone, 0.3 mL; 50/PK RK-07904-34 Mfr # 701357
COMPARE
Silicone 384 0.3
$1,059.37CAD / Pkg of 50