This product is now discontinued, View Related Products

Brady Labelmark 5 Laboratory Labeling Software

Quickly create, view, and print laboratory labels
  • Import, edit, and view data for multiple labels at once
This labeling software can integrate data sources, label designs, and batch printing into one cohesive system — a time-saving solution for operational label setup, management, and execution. With single-click importing and serialization, an interactive custom label creator, and a complete integration with MS Office and AutoCAD applications, this labeing software is a user-friendly label design and printing software for high-performance labeling. Print labels simultaneously on multiple printers for fast label kitting. Design fixed text labels for quick printing or free-form labels for custom label design.


System requirements: Windows® Server 2003; Windows Server 2008; Windows Vista with SP1; Windows XP with SP3; 1GHz Pentium processor or higher; 512 MB or higher RAM;
Up to 500 MB of available space may be required; DVD Drive; 1024 x 768 high color, 32-bit or better display.

$31.68 - $375.71CAD / Each

9 variations of this product are available.

See All Product Options Below
9 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Brady PTL-82-499 TLS 2200/TLS PC Link Labels for 0.5- 1.5 mL Vials, White, Nylon RK-65543-18 Mfr # PTL-82-499
COMPARE
TLS 2200/TLS PC Link Labels for 0.5- 1.5 mL Vials, White, Nylon
$63.72CAD / Each
Brady PTL-98-499 Label for 0.5 to 1.5 mL tube/vial tops and sides RK-65543-22 Mfr # PTL-98-499
COMPARE
Label for 0.5 to 1.5 mL tube/vial tops and sides
$125.97CAD / Each
Brady PTL-83-499 TLS 2200/TLS PC Link Labels for 2-50 mL Vials, White, Nylon RK-65543-24 Mfr # PTL-83-499
COMPARE
TLS 2200/TLS PC Link Labels for 2-50 mL Vials, White, Nylon
$62.99CAD / Each
Brady PTL-97-488 Label for microscope slides, 22.9 x 22.9 mm RK-65543-30 Mfr # PTL-97-488
COMPARE
Label for microscope slides, 22.9 x 22.9 mm
$53.04CAD / Each
Brady PTL-86-461 Labels for 1 to 2 mL centrifuges, cryo vials, Eppendorf, PCR tubes - roll of 100 RK-65543-34 Mfr # PTL-86-461
COMPARE
Labels for 1 to 2 mL centrifuges, cryo vials, Eppendorf, PCR tubes - roll of 100
$47.52CAD / Each
Brady PTL-74-461 Labels for 1 to 6 mL centrifuges, cryo vials, Eppendorf, PCR tubes, roll of 100 RK-65543-36 Mfr # PTL-74-461
COMPARE
Labels for 1 to 6 mL centrifuges, cryo vials, Eppendorf, PCR tubes, roll of 100
$31.68CAD / Each
Brady PTL-112-490 BMP-71 Labels for 0.5-0.7 mL/Eppendorf/PCR/Tubes; 500/Roll RK-65543-38 Mfr # PTL-112-490
COMPARE
BMP-71 Labels for 0.5-0.7 mL/Eppendorf/PCR/Tubes; 500/Roll
$78.09CAD / Each
Brady PTL-80-488 Labels for microscope slides, 19 x 22.9 mm, roll of 250 RK-65543-40 Mfr # PTL-80-488
COMPARE
Labels for microscope slides, 19 x 22.9 mm, roll of 250
$49.36CAD / Each
Brady LM5STDCD Labelmark 5 Laboratory Labeling Software RK-65543-60 Mfr # LM5STDCD
COMPARE
Labelmark 5 Laboratory Labeling Software
$375.71CAD / Each
Showing 1- 9 of 9

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat