Justrite Antistatic Bonding Wire with Dual Hand Clamp
Prevent sparks from static electricity
  • Standard wire lengths are 3 ft (1 m) and 10-foot (3-m)
Bonding wires prevent sparks from static electricity generated by movement and flow of flammable liquids. Bonding between containers during liquid transfer and connecting drums to an earth ground is the safest method for transferring flammable liquids.
During waste collection operations, use one antistatic wire to bond from a safety container to a receiving vessel such as a funnel, then properly ground by attaching a second antistatic wire to the drum and then to an earth ground.
$211.43CAD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Justrite 08499 Antistatic Wire for Bonding and Grounding w/Dual Hand Clamps, 3' RK-09417-10 Mfr # 08499
COMPARE
Bonding wire, two hand clamps with 1\" jaws
$211.43CAD / Each