This product is now discontinued, View Related Products
Boekel Scientific™ Touch Screen CO2 Incubator
Advanced color touch screen for custom temperature and CO2 programs
  • Quickly download data via USB or Ethernet connections
  • Easily add humidity to the chamber with included humidity pan
  • Constructed with a polished stainless steel interior
This touch screen CO2 incubator provides easy viewing of alarms, current and set temperatures, CO2 level and set point, and humidity levels. The 4.3” color display can also show temperature, humidity and CO2 graphs which can then be exported via USB and Ethernet connections. Setpoint ramp programming can be used to specify multistep ramp profiles, controlling carbon dioxide, temperature, and outlet conditions over specified time periods.
Constructed with heated double doors and an air jacketed resistive heater array, this incubator maintains a controlled environment, ideal for the protection of cell cultures, bacteria cultures or tissue cultures. The double door design features a clear tempered glass viewing door, for viewing your samples without disrupting the chamber environment.
$10,048.52 - $10,520.00CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Capacity (cu ft) Max Temperature (° C) Power (VAC)
Availability Pricing
Boekel 155000 Touch Screen CO2 Incubator, 120V RK-01100-01 Mfr # 155000
COMPARE
4 90 115
$10,048.52CAD / Each
Boekel 155000-2 Touch Screen CO2 Incubator, 230V RK-01100-02 Mfr # 155000-2
COMPARE
4 90 230
$10,520.00CAD / Each