This product is now discontinued, View Related Products
Boekel Scientific Orbitron V 3-D Orbital Wave Motion Shaker
Heavy-duty tray can hold up to 20 lb
  • Skid-free rubber mat keeps vessels in the proper position
  • Run continuously or up to 12 hours with built in timer
This shaker features smooth three-dimensional orbital wave motion to mix through any medium. From gentle to vigorous, the adjustable tray and variable speed allows you to get the proper agitation you need for your application. The tray can be adjusted from 0 to 12° in seconds with a single set screw. Adjust the speed from 5 to 120 rpm with a simple turn of the dial. Add additional tray (sold separately) to stack your samples.
$3,519.93 - $3,717.36CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Power (VAC)
Availability Pricing
Boekel Scientific 281111 Scientific Orbitron V 3-D Orbital Wave Motion Shaker, 115V RK-51705-02 Mfr # 281111
COMPARE
115
$3,519.93CAD / Each
Boekel Scientific 281111-2 Scientific Orbitron V 3-D Orbital Wave Motion Shaker, 230V RK-51705-04 Mfr # 281111-2
COMPARE
230
$3,717.36CAD / Each