High-Performance Exhaust Blowers
Ready removal of harmful fumes – anywhere in the workplace!
Choose the exhaust blower suited to your need. Order optional Y-fitting to adapt two extractor arms to a blower. Optional silencer for ½ and ¾ hp blowers reduces noise by 3 to 4 dBA. Alternative blowers are available for excessively corrosive fumes. Contact Cole-Parmer Applications for blower selection assistance and guidance regarding adaptive fittings and hosing connections.
Not recommended for highly corrosive exhaust streams.
What’s included: inlet and outlet collars, wall bracket.
$1,195.66CAD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Blower Type Exhaust Volume (CFM) Power (VAC)
Availability Pricing
Air Impurities Removal Systems Exhaust Blower, 350 cfm, 350 Watt, 115 VAC RK-33704-75 Mfr # B-982-1A
COMPARE
Standard 350 115
$1,195.66CAD / Each