This product is now discontinued, View Related Products
Biowarfare Agent Detection Devices (BADD)
Quickly determine biological threat credibility
  • Results in 3- 30 minutes
  • Easy to read – two lines for positive; one line for negative
  • High Sensitivity and specificity
Accurately detect small, yet high threat amounts of biowarfare agents-without extensive sample preparation or additional expensive equipment. Test powder or liquid samples; kits are simple to use, results are quick and easy to read. Biohazard disposal bags and chain of custody labels are included for proper control and disposal or transport of the sample and test. Order simulation devices below for training purposes.
What’s included: 5" x 7" pouch includes all material to prepare and test a sample as well as package it for disposal or transport after testing.
$714.59 - $3,027.82CAD / Box of 10
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Number of Tests Included Test Type
Availability Pricing
COMPARE
10 Anthrax
$926.76CAD / Box of 10
COMPARE
10 Botulinum Toxin
$906.13CAD / Box of 10
COMPARE
10 Ricin Toxin
$926.76CAD / Box of 10
COMPARE
10
$714.59CAD / Box of 10
COMPARE
30 Anthrax, Botulinum Toxin, Ricin
$3,027.82CAD / Box of 30
COMPARE
Salmonella
$884.03CAD / Box