This product is now discontinued, View Related Products
BINDER KB Series Refrigerated Incubators
Precision control for maintaining specimen quality at low temperatures.
  • Incubation from -10°C to 100°C.
  • Rapid temperature recovery rates.
  • Disenfection routine up to 100°C.
All models feature APT.line® electronic controllers and preheating chambers, offering precise consistent temperature performance and industry leading recovery times after door openings. Specially designed two-point door closure system and thermal insulation provided added protection from changing environmental conditioins. The inner glass door seals hermetically to allow users to monitor cultures without influencing chamber conditions. Units also feature independent adjustable temperature safety devices with visual alarm. RS-422 interface for standard software guarantees seamless documentation of all testing parameters.
$7,985.77 - $23,250.09CAD / Each
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
Compareitem specifications
Capacity (cu ft) Power (VAC)
Availability Pricing
Binder Kb Series Refrigerated Incubator 0.7 Cu. Ft. 115V RK-39071-00 Mfr # 9020-0113
COMPARE
0.7 115
$7,985.77CAD / Each
Binder Kb Series Refrigerated Incubator 0.7 Cu. Ft. 230V RK-39071-02 Mfr # 9020-0112
COMPARE
230
$8,214.15CAD / Each
Binder Kb Series Refrigerated Incubator 1.9 Cu. Ft. 115V RK-39071-04 Mfr # 9020-0115
COMPARE
115
$8,914.01CAD / Each
Binder Kb Series Refrigerated Incubator 1.9 Cu. Ft. 230V RK-39071-06 Mfr # 9020-0114
COMPARE
230
$8,516.19CAD / Each
Binder Kb Series Refrigerated Incubator 4.1 Cu. Ft. 115V RK-39071-08 Mfr # 9020-0253
COMPARE
4.1 115
$12,114.21CAD / Each
Binder Kb Series Refrigerated Incubator 4.1 Cu. Ft. 230V RK-39071-10 Mfr # 9020-0116
COMPARE
230
$10,652.61CAD / Each
Binder Kb Series Refrigerated Incubator 8.6 Cu. Ft. 115V RK-39071-12 Mfr # 9020-0119
COMPARE
8.6 115
$12,155.47CAD / Each
Binder Kb Series Refrigerated Incubator 8.6 Cu. Ft. 230V RK-39071-14 Mfr # 9020-0118
COMPARE
8.6 230
$12,155.47CAD / Each
Binder Kb Series Refrigerated Incubator 14.3 Cu. Ft. 115V RK-39071-16 Mfr # SUB:9020-0179
COMPARE
115
$17,106.06CAD / Each
Binder Kb Series Refrigerated Incubator 14.3 Cu. Ft. 230V RK-39071-18 Mfr # 9020-0178
COMPARE
230
$17,106.06CAD / Each
Binder Kb Series Refrigerated Incubator 25.1 Cu. Ft. 115V RK-39071-20 Mfr # 9020-0167
COMPARE
115
$23,250.09CAD / Each
Binder Kb Series Refrigerated Incubator 25.1 Cu. Ft. 230V RK-39071-22 Mfr # 9020-0111
COMPARE
230
$23,250.09CAD / Each
Showing 1- 12 of 12