Best Viton Gloves
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
Gloves made of Viton® fluoroelastomer; provide superior resistance to PCBs, chlorinated and aromatic solvents, and vapors. Smooth, flexible, unlined gloves with a rolled cuff are injection-molded for consistent thickness throughout.
$218.06 - $275.52CAD / Pair of 1
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Size
Availability Pricing
Showa Best 892-8 Viton Gloves, small, 12-mil thickness, 1 pair RK-86288-20 Mfr # 892-8
COMPARE
12 Small
$218.06CAD / Pair of 1
Showa Best 892-9 Viton Gloves, medium, 12-mil thickness, 1 pair RK-86288-21 Mfr # 892-9
COMPARE
12 Medium
$241.64CAD / Pair of 1
Showa Best 892-10 Viton Gloves, large, 12-mil thickness, 1 pair RK-86288-22 Mfr # 892-10
COMPARE
12 Large
$241.64CAD / Pair of 1
Showa Best 892-11 Viton Gloves, extra-large, 12-mil thickness, 1 pair RK-86288-23 Mfr # 892-11
COMPARE
12 X-Large
$241.64CAD / Pair of 1
Showa Best 890-09 Viton Gloves, large, 28-mil thickness, 1 pair RK-86288-32 Mfr # 890-09
COMPARE
14 Large
$275.52CAD / Pair of 1
Showa Best 890-10 Viton Gloves, extra-large, 28-mil thickness, 1 pair RK-86288-33 Mfr # 890-10
COMPARE
14 X-Large
$275.52CAD / Pair of 1