This product is now discontinued, View Related Products
SHOWA N-DEX Laboratory Grade Nitrile Gloves
Reduce hand fatigue by keeping your hands cool and dry
  • Nonlatex—made of soft 100% nitrile
  • Three times the puncture-resistance of natural rubber or vinyl
  • Superior tactile-sensitivity, comfort, and dexterity
  • Lightweight and disposable
N-DEX® gloves are made from a special low-modulus, nonlatex, soft nitrile formulation. They offer greater tactile-sensitivity, comfort, dexterity, and puncture-resistance than latex gloves. All N-DEX® nitrile gloves are ambidextrous and blue in color.
General and Laboratory Grade N-DEX® Gloves are ideal for use anywhere you need to protect your hands from the environment. Box of 100.
$31.68 - $38.90CAD / Pkg of 100
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Length (in) Powdered Size
Availability Pricing
SHOWA 7005S Laboratory Grade Gloves, low-powder, small RK-86287-70 Mfr # 7005S
COMPARE
9 1/2 Yes Small
$31.68CAD / Pkg of 100
SHOWA 7005M Laboratory Grade Gloves, low-powder, medium RK-86287-71 Mfr # 7005M
COMPARE
9 1/2 Yes Medium
$32.05CAD / Pkg of 100
SHOWA 7005L Laboratory Grade Gloves, low-powder, large RK-86287-72 Mfr # 7005L
COMPARE
9 1/2 Yes Large
$32.05CAD / Pkg of 100
SHOWA 7005XL Laboratory Grade Gloves, low-powder, X-large RK-86287-73 Mfr # 7005XL
COMPARE
9 1/2 Yes X-Large
$31.68CAD / Pkg of 100
SHOWA 7005PFS Laboratory Grade Gloves, powder-free, small RK-86287-80 Mfr # 7005PFS
COMPARE
9 1/2 No Small
$31.68CAD / Pkg of 100
SHOWA 7005PFM Laboratory Grade Gloves, powder-free, medium RK-86287-81 Mfr # 7005PFM
COMPARE
9 1/2 No Medium
$32.05CAD / Pkg of 100
SHOWA 7005PFL Laboratory Grade Gloves, powder-free, large RK-86287-82 Mfr # 7005PFL
COMPARE
9 1/2 No Large
$31.68CAD / Pkg of 100
SHOWA 7005PFXL Laboratory Grade Gloves, powder-free, X-large RK-86287-83 Mfr # 7005PFXL
COMPARE
9 1/2 No X-Large
$38.90CAD / Pkg of 100
Showing 1- 8 of 8