This product is now discontinued, View Related Products
Beijer iX Series Rugged Human Machine Interfaces
Operate in extreme temperatures and other harsh or dangerous environments
  • Extensive certifications ensure function despite heat, cold, water, vibration, or shock
  • Galvanic isolation protects against both bursts and surges in electricity
  • IP66/NEMA 4X front-panel enclosure resists snow, rain, high-pressure washdown, dirt, and dust
  • Vibration/shock: 4 g RMS sweep sine (vibration), 40 g 11 ms half-sine (shock)
Ideally suited to oil and gas, marine, and water/wastewater industries, and solar array or engine room controls, these human machine interfaces (HMI) feature specialized components to increase durability and undergo rigorous testing to ensure operation under the toughest conditions. Hazardous environment certifications include UL Class I Div 2, ATEX Zone 2, and IECEx Zone 2. Marine environment certifications include DNV, ABS, GL, LR, KR, and EN60945.
Both the serial and the CAN ports are galvanically isolated, embedded NAND flash memory is designed for extreme temperatures and extended write cycles, and touch screen is composed of chemically strengthened glass for longer service life. The panels accept 12 or 24 VDC, so converters are unnecessary.
Optional features:
High-performance models (45004-22, -23, -25, -26) feature an enhanced processor. The standard is 1.0 GHz, while the high performance version is 1.6 GHz.
High-brightness models (45004-22, -23, -25, -26, -28, -29) offer a brighter backlight suitable for easy reading in direct sunlight. Both the standard and the high-brightness models are dimmable for low-light conditions.
CAN module (45004-24 to -26, -29) allows HMI to interface with controllers using CAN communication channels.
Glossy display (45004-22 and -25) is best suited for panels installed in low-light or diffuse-light conditions. The reflective coating provides better contrast and color saturation than the standard matte coating; however, it also suffers from glare in bright rooms and on sunny days.
$884.03 - $6,294.32CAD / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Beijer Electronics iX Series  Rugged Human Machine Interface, 15" Matte Display, 12-24 VDC RK-45004-21 Mfr # IXT15BR/4521-640003701
COMPARE
Rugged Human Machine Interface, 15\" Matte Display, 12-24 VDC
$5,417.66CAD / Each
Beijer Electronics iX Series  Rugged HMI, 15" Extra-Bright Glossy, High Performance, 12-24 VDC RK-45004-22 Mfr # IXT15BR/4522-640004001
COMPARE
Rugged HMI, 15\" Extra-Bright Glossy, High Performance, 12-24 VDC
$6,054.90CAD / Each
Beijer Electronics iX Series  Rugged HMI, 15" Extra-Bright Matte, High Performance, 12-24 VDC RK-45004-23 Mfr # IXT15BR/4523-640004501
COMPARE
Rugged HMI, 15\" Extra-Bright Matte, High Performance, 12-24 VDC
$6,054.90CAD / Each
Beijer Electronics iX Series  Rugged HMI, 15" Matte Display, CAN Module, 12-24 VDC RK-45004-24 Mfr # IXT15BR/4524-640004401
COMPARE
Rugged HMI, 15\" Matte Display, CAN Module, 12-24 VDC
$5,657.08CAD / Each
Beijer Electronics iX Series  Rugged HMI, 15" Extra-Bright Glossy, High Performance, CAN, 12-24 VDC RK-45004-25 Mfr # IXT15BR/4525-640004801
COMPARE
Rugged HMI, 15\" Extra-Bright Glossy, High Performance, CAN, 12-24 VDC
$6,294.32CAD / Each
Beijer Electronics iX Series  Rugged HMI, 15" Extra-Bright Matte, High Performance, CAN, 12-24 VDC RK-45004-26 Mfr # IXT15BR/4526-640004601
COMPARE
Rugged HMI, 15\" Extra-Bright Matte, High Performance, CAN, 12-24 VDC
$6,294.32CAD / Each
Beijer Electronics iX Series  Rugged Human Machine Interface, 7" Matte Display, 12-24 VDC RK-45004-27 Mfr # IXT7BR/4712R-640007101
COMPARE
Rugged Human Machine Interface, 7\" Matte Display, 12-24 VDC
$4,424.59CAD / Each
Beijer Electronics iX Series  Rugged HMI, 7" Extra-Bright Matte Display, 12-24 VDC RK-45004-28 Mfr # IXT7BR/4714R-640007301
COMPARE
Rugged HMI, 7\" Extra-Bright Matte Display, 12-24 VDC
$3,186.94CAD / Each
Beijer Electronics iX Series  Rugged HMI, 7" Extra-Bright Matte Display, CAN, 12-24 VDC RK-45004-29 Mfr # IXT7BR/4717R-640007601
COMPARE
Rugged HMI, 7\" Extra-Bright Matte Display, CAN, 12-24 VDC
$3,426.37CAD / Each
Beijer Electronics iX Developer Software Human Machine Interface RK-68516-50 Mfr # iX Developer Software
COMPARE
Human Machine Interface
$884.03CAD / Each
Showing 1- 10 of 10