This product is now discontinued, View Related Products
Balmac Vibration Transmitters
FEATURES
  • Solid-state circuitry
  • Two-wire 4-20 mA signal format
  • Same wiring for both power and vibration signal
  • Economical for continuous monitoring of all machines
  • Connects in series with DC power supply and measurement devices
  • Works with single-loop monitors
  • Rugged NEMA 4 weatherproof case
APPLICATIONS
  • Fans, Motors, Pumps and Compressors
  • Controlling Vibrating Conveyors
  • General Purpose Machinery
  • Hazardous Environments
$707.23CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Max Vibration Range - Velocity (in/sec)
Availability Pricing
Balmac Inc. 191-5 Vibration Transmitter, 0-5" /Sec RK-98853-11 Mfr # 191-5
COMPARE
5
$707.23CAD / Each
Balmac Inc. 191-1 Vibration Transmitter, 0-1 In/Sec RK-98853-14 Mfr # 191-1
COMPARE
1
$707.23CAD / Each
Balmac Inc. 191-2 Vibration Transmitter, 0 to In./Sec RK-98853-15 Mfr # 191-2
COMPARE
2
$707.23CAD / Each