This product is now discontinued, View Related Products
Balmac Digital Vibration Kits
Measure vibration velocity, displacement, and acceleration
  • Measures and stores 250 overall vibration amplitude measurements
  • Industrial 100 mV/g ICP accelerometer
  • Automatic amplitude scaling
  • Hand-held, portable, battery-operated
  • Easy-to-read back-lit graphic display
  • Magnetic base
  • 7" probe extension
  • Auto power off
$2,320.59CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Vibration Meter, English
$2,320.59CAD / Each
Balmac Inc. 230M Vibration Meter with Metric units of measure RK-65700-12 Mfr # 230M
COMPARE
Vibration Meter, Metric
$2,320.59CAD / Each