This product is now discontinued, View Related Products
Bürkle Wide-Mouth Sampling Bottles with Tamper-Evident Safety Cap
Maintain authenticity and security with sampling bottle and seal
  • Uniquely identify sample with tamper-evident seal sold separately
  • Constructed of durable HDPE bottles
  • Leakproof cap design for safe handling or shipping
These wide-mouth sampling bottles are ideal for sample storage or sending samples as they feature sturdy HDPE construction and leakproof cap. The included cap features an eyelet that allows the sampling seal, sold separately, to secure the cap. The seal is secured with a metal spring, allowing for one-way tightening and security against manipulation. The seal also features clear identification via a 9-digit number printed on the seal.
$6.26 - $14.37CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Capacity (mL) Neck Inside Diameter (ID) (in) Overall Height (in)
Availability Pricing
Burkle 0322-0250 Sampling bottle with tamper-proof safety cap, HDPE, 250 mL RK-06040-17 Mfr # 0322-0250
COMPARE
250 1 1/25 4 45/64
$6.26CAD / Each
Burkle 0322-0500 Sampling bottle with tamper-proof safety cap, HDPE, 500 mL RK-06040-18 Mfr # 0322-0500
COMPARE
500 1 9/16 5 51/64
$6.26CAD / Each
Burkle 0322-1000 Sampling bottle with tamper-proof safety cap, HDPE, 1000 mL RK-06040-19 Mfr # 0322-1000
COMPARE
1000 2 7 19/64
$12.52CAD / Each
Burkle 0322-2000 Sampling bottle with tamper-proof safety cap, HDPE, 2000 mL RK-06040-20 Mfr # 0322-2000
COMPARE
2000 2 9
$14.37CAD / Each