This product is now discontinued, View Related Products
Bürkle Stainless Steel Uno Samplers for Powders and Granulates
Reliably sample various bulk goods, powders, and granulates for different sampling tasks
  • Efficient sampling made possible with the smallest gap possible between the inner and outer tube
  • Constructed of ultrapure materials for a durable design
These maintenance-free samplers allow for collection from the finest powders to large grains. They are suitable for sampling from all layers of the container, providing an accurate representation of the contents. These samplers come in different lengths, with single or multiple chambers and openings, and made of different material options for all different sampling methods. The 55 cm long samplers are suitable for sacks; the 85 cm long samplers are ideal for barrels, tubs, and mixed containers; and the 150 cm long samplers are appropriate for big bags, silos, and tanks.
The uno samplers feature a single closed sample chamber and one chamber opening for point sampling at a predetermined depth. The outer tube is made of 316L stainless steel and the sample chamber is made of PTFE, making these chemically resistant and suitable for steam sterilization up to 250°F (121°C). Color-coded identification tags offer quick identification and prevent sample mix-ups.
$1,127.14 - $1,824.79CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Length (in) Volume (mL) Description
Availability Pricing
Burkle 5316-1055 Sampling Lance, SS/PTFE, one chamber; 17 mL capacity RK-06041-37 Mfr # 5316-1055
COMPARE
21.7 17 Single chamber sampling lance, SS/PTFE, one chamber, 55 cm length, 430 mm insertion depth; 17 mL capacity
$1,127.14CAD / Each
Burkle 5316-1085 Sampling Lance, SS/PTFE, one chamber; 17 mL capacity RK-06041-38 Mfr # 5316-1085
COMPARE
33.5 17 Single chamber sampling lance, SS/PTFE, one chamber, 85 cm length, 710 mm insertion depth; 17 mL capacity
$1,261.22CAD / Each
Burkle 5316-1150 Sampling Lance, SS/PTFE, one chamber; 17 mL capacity RK-06041-39 Mfr # 5316-1150
COMPARE
59 17 Single chamber sampling lance, SS/PTFE, one chamber, 150 cm length, 1355 mm insertion depth; 17 mL capacity
$1,824.79CAD / Each