This product is now discontinued, View Related Products
Bürkle Stainless Steel Single Chamber Samplers for Powders and Granulates
Reliably sample various bulk goods, powders, and granulates for different sampling tasks
  • Efficient sampling made possible with the smallest gap possible between the inner and outer tube
  • Constructed of ultrapure materials for a durable design
These maintenance-free samplers allow for collection from the finest powders to large grains. They are suitable for sampling from all layers of the container, providing an accurate representation of the contents. These samplers come in different lengths, with single or multiple chambers and openings, and made of different material options for all different sampling methods. The 55 cm long samplers are suitable for sacks; the 85 cm long samplers are ideal for barrels, tubs, and mixed containers; and the 150 cm long samplers are appropriate for big bags, silos, and tanks.
Single-chamber samplers feature an open inner tube with no separate chambers, allowing for sampling large volumes. These samplers feature either 3 or 5 openings for sample collection of all layers. These are available in either aluminum or 316L stainless steel options and are suitable for steam sterilization up to 250°F (121°C).
$511.41 - $2,080.43CAD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Material Volume (mL) Description
Availability Pricing
Burkle 5316-2055 Sampling Lance, Stainless Steel, 3 openings, one chamber; 70 mL capacity RK-06041-40 Mfr # 5316-2055
COMPARE
316L Stainless Steel 70 All layer sampling lance, SS, 3 openings, one chamber, 55 cm length, 430 mm insertion depth; 70 mL capacity
$1,516.86CAD / Each
Burkle 5316-2085 Sampling Lance, Stainless Steel, 3 openings, one chamber; 130 mL capacity RK-06041-41 Mfr # 5316-2085
COMPARE
316L Stainless Steel 130 All layer sampling lance, SS, 3 openings, one chamber, 85 cm length, 710 mm insertion depth; 130 mL capacity
$1,843.21CAD / Each
Burkle 5316-2150 Sampling Lance, Stainless Steel, 5 openings, one chamber; 260 mL capacity RK-06041-42 Mfr # 5316-2150
COMPARE
316L Stainless Steel 260 All layer sampling lance, SS, 5 openings, one chamber, 150 cm length, 1355 mm insertion depth; 150 mL capacity
$2,080.43CAD / Each
Burkle 5317-2055 Sampling Lance, Aluminum, 3 openings, one chamber; 70 mL capacity RK-06041-51 Mfr # 5317-2055
COMPARE
Aluminium 70 All layer sampling lance, aluminum, 3 openings, one chamber, 55 cm length, 430 mm insertion depth; 70 mL capacity
$511.41CAD / Each
Burkle 5317-2085 Sampling Lance, Aluminum, 3 openings, one chamber; 130 mL capacity RK-06041-52 Mfr # 5317-2085
COMPARE
Aluminium 130 All layer sampling lance, aluminum, 3 openings, one chamber, 85 cm length, 710 mm insertion depth; 130 mL capacity
$573.15CAD / Each
Burkle 5317-2150 Sampling Lance, Aluminum, 5 openings, one chamber; 150 mL capacity RK-06041-53 Mfr # 5317-2150
COMPARE
Aluminium 260 All layer sampling lance, aluminum, 5 openings, one chamber, 150 cm length , 1355 mm insertion depth; 150 mL capacity
$727.85CAD / Each