This product is now discontinued, View Related Products
Bürkle Stainless Steel Powder Sampling Lances
Quickly sample powders and granules from bags or casks
  • Constructed of 316L stainless steel—ideal for food and pharma applications
  • Suitable for steam sterilization up to 250°F (121°C)
  • No-hassle maintenance-free sampler
Sampler model 06041-62 is a universal powder and granule sampler initially developed for milk powder samples, and conforms to the standards of DIN and IDF (International Dairy Federation). This unit features a semi-circular profile lance with an insertion depth of 15.2" (38.5 cm) and sample volume of 150 mL.
Sampler models 06041-85 and -86 feature a slender, pointed tip that can be inserted easily and deeply to take a 50-mL sample. One-piece construction without edges or groves offers easy cleaning. The sample can be filled directly into the sampling bag from the outlet end. Both models come with a clamp and 100 polyethylene bags (120 x 170 mm).
$446.29 - $1,128.25CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Diameter (in) Length (in) Volume (mL)
Availability Pricing
Burkle 5321-0300 Powder/granular sampling lance, SS, 385 mm insertion depth; 150 mL RK-06041-62 Mfr # 5321-0300
COMPARE
1.1 18.9 150
$446.29CAD / Each
Burkle 5338-0410 Tampered conical lance sampler with 50 mL sample bags, SS, 410 mm x 25 mm, meets ISTA standards RK-06041-85 Mfr # 5338-0410
COMPARE
1 16.1 50
$841.31CAD / Each
Burkle 5338-1100 Tampered conical lance sampler with 50 mL sample bags, SS, 1100 mm x 25 mm, meets ISTA standards RK-06041-86 Mfr # 5338-1100
COMPARE
1 43.3 50
$1,128.25CAD / Each