This product is now discontinued, View Related Products
Bürkle Stainless Steel Pointed Samplers for Bulk Goods
Easily sample powders or granules at targeted depths
  • Constructed of 316L stainless steel
  • Quickly empty the sample without having to disassemble the sampler
  • Autoclavable up to 121°C for 20 minutes
These powder and granulate samplers are designed to take a target point sample at the position within the sample material that has precisely been determined. The sampler is closed when it is inserted into the sample material, opened when it reaches the target point, and closed again once the sample is obtained.
The pointed stainless steel samplers are available in two chamber volume options: 60 or 200 mL. The sample can be emptied directly from the sampler into a container, without disassembling the sampler, which is useful when obtaining a mixed sample.
$1,665.67 - $1,690.72CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Diameter (in) Length (in) Volume (mL)
Availability Pricing
Burkle 5360-0600 Pointed-plunger lance sampler, SS, 100 cm x 32 mm; 60 mL RK-06042-17 Mfr # 5360-0600
COMPARE
1.3 39.4 60
$1,665.67CAD / Each
Burkle 5360-1000 Pointed-plunger lance sampler, SS, 104.5 cm x 32 mm; 200 mL RK-06042-18 Mfr # 5360-1000
COMPARE
1.3 41.1 200
$1,690.72CAD / Each