This product is now discontinued, View Related Products
Bürkle Stainless Steel Mini ViscoSampler Tips
Interchangeable tips for the Bürkle Stainless Steel Mini ViscoSamplers
  • Ideal for pharmaceutical and cosmetics applications with the easy to clean design
  • Suitable for steam sterilization up to 250°F (121°C)
There are three removable tip sizes to choose from, with 2 mm to 6 mm holes, depending on the viscosity of the samples. The sample is drawn into the pump rod and transported directly to the sampling container. Sampler is sold separately as part numbers 06041-92 through -94.
$112.35CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Material Description
Availability Pricing
Burkle 5341-1002 Head for small viscous substance sampling rod, SS; 2 mm RK-06041-89 Mfr # 5341-1002
COMPARE
316L Stainless Steel Burkle, Inc. 5341-1002 Head for small viscous substance sampling rod, SS; 2 mm
$112.35CAD / Each
Burkle 5341-1004 Head for small viscous substance sampling rod, SS; 4 mm RK-06041-90 Mfr # 5341-1004
COMPARE
316L Stainless Steel Burkle, Inc. 5341-1004 Head for small viscous substance sampling rod, SS; 4 mm
$112.35CAD / Each
Burkle 5341-1006 Head for small viscous substance sampling rod, SS; 6 mm RK-06041-91 Mfr # 5341-1006
COMPARE
316L Stainless Steel Burkle, Inc. 5341-1006 Head for small viscous substance sampling rod, SS; 6 mm
$112.35CAD / Each