This product is now discontinued, View Related Products
Bürkle Piercing Core Samplers with Collection Bottle for Bulk Goods and Granulates
Quickly collect samples with this all-in-one sampler and collection bottle
  • Easily sample directly from sacks or bags with the durable piercing tip
  • Suitable for steam sterilization up to 121°C
  • Maintenance free with removable tip for easy cleaning
These core samplers are suitable for sampling powders or granulates directly from sacks or bags. The samples collected fill directly into the attached 250-mL LDPE sampling bottle. These samplers have no undercuts or edges in which residue can accumulate, making them ideal for quality control applications.
Samplers have an insertion depth of 30 cm and a 2.5 cm diameter. The stainless steel sampler is approved for use with foodstuff, while the transparent polypropylene sampler is ideal for use with aggressive chemicals. Both samplers correspond to ISTA standards for seed lengths of 10 mm (wheat and oats) and 5 mm (flax and vetches).
The collection bottle screws directly onto the threads of the core sampler, and has a 31-mm neck inner diameter and 40-mm thread diameter.
$358.03 - $569.02CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Diameter (in) Material Volume (mL)
Availability Pricing
Burkle 5322-0300 Core sampler, PP, 30 cm x 25 mm, meets ISTA standards; 75 mL RK-06041-63 Mfr # 5322-0300
COMPARE
1 Stainless Steel 75
$358.03CAD / Each
Burkle 5322-1300 Core sampler, SS, 30 cm x 25 mm, meets ISTA standards; 75 mL RK-06041-64 Mfr # 5322-1300
COMPARE
1 Polypropylene 75
$569.02CAD / Each