This product is now discontinued, View Related Products
Bürkle MicroDiso Samplers
Target-point, small-volume, disposable samplers
  • 10-mL sample volume for critical powders and granules
  • Acquire specimens from 500 or 1000 mm depths
  • 21-mm diameter fits in tight spaces
  • Disposable, all-HDPE construction to reduce cross-contamination
  • Pointed-tip plunger aids in full sample expulsion
  • Individually-wrapped and cleanroom packaged for ultimate cleanliness
  • Sterile models have been decontaminated via gamma irradiation
These disposable HDPE samplers are ideal for collecting targeted-depth, powder/small granulated samples. Available non-sterilized or pre-sterilized via gamma irradiation.
$345.07 - $520.99CAD / Pkg of 10
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Length (cm) Sterile
Availability Pricing
Burkle 5393-2240 Small disposable pointed-plunger lance sampler, HDPE, 500 mm x 21 mm; 10 mL RK-06043-06 Mfr # 5393-2240
COMPARE
50 No
$345.07CAD / Pkg of 10
Burkle 5393-2241 Small disposable pointed-plunger lance sampler, HDPE, 500 mm x 21 mm, sterile; 10 mL RK-06043-07 Mfr # 5393-2241
COMPARE
50 Yes
$407.69CAD / Pkg of 10
Burkle 5393-2250 Small disposable pointed-plunger lance sampler, HDPE, 1000 mm x 21 mm; 10 mL RK-06043-08 Mfr # 5393-2250
COMPARE
100 No
$410.93CAD / Pkg of 10
Burkle 5393-2251 Small disposable pointed-plunger lance sampler, HDPE, 1000 mm x 21 mm, sterile; 10 mL RK-06043-09 Mfr # 5393-2251
COMPARE
100 Yes
$520.99CAD / Pkg of 10