This product is now discontinued, View Related Products
Bürkle Disposable Target Sampling Lances for Powders and Small Granulates
Quickly and easily take a sample at a specified point within the vessel
  • Manufactured in a cleanroom ISO Class 7 environment and individually packaged
  • Certified FDA 21 CFR 5 compliant for food applications
These powder and granulate TargetDispo samplers are designed to take a target point sample at an exact position within the vessel. The sampler is closed when it is inserted into the sample material, opened when it reaches the targeted point, and closed again once the sample is obtained. The sample can then be emptied through the open handle. Choose from two capacity options in either sterile or nonsterile configurations. The sterilized options are sterilized via gamma irradiation. The diameter of the sampler is 2.5 cm and the diameter of the handle is 1.4 cm. HDPE construction.
$428.76 - $599.38CAD / Pkg of 20
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Length (in) Volume (mL) Description
Availability Pricing
Burkle 5393-4460 Target Closed-tip Lance Sampler, HDPE, 500 mm x 21 mm; 500 mL RK-06043-24 Mfr # 5393-4460
COMPARE
19.7 100 Target closed-tip lance sampler, HDPE, 500 mm x 21 mm; 500 mL
$428.76CAD / Pkg of 20
Burkle 5393-4461 Target Closed-tip Lance Sampler, HDPE, 500 mm x 21 mm, Sterile; 500 mL RK-06043-25 Mfr # 5393-4461
COMPARE
19.7 100 Target closed-tip lance sampler, HDPE, 500 mm x 21 mm, sterile; 500 mL
$494.76CAD / Pkg of 20
Burkle 5393-4470 Target Closed-tip Lance Sampler, HDPE, 1000 mm x 21 mm; 1000 mL RK-06043-26 Mfr # 5393-4470
COMPARE
39.4 200 Target closed-tip lance sampler, HDPE, 1000 mm x 21 mm; 1000 mL
$534.25CAD / Pkg of 20
Burkle 5393-4471 Target Closed-tip Lance Sampler, HDPE, 1000 mm x 21 mm, Sterile; 1000 mL RK-06043-27 Mfr # 5393-4471
COMPARE
39.4 200 Target closed-tip lance sampler, HDPE, 1000 mm x 21 mm, sterile; 1000 mL
$599.38CAD / Pkg of 20