This product is now discontinued, View Related Products
Bürkle Disposable Biopolyethylene Food-Grade Sampling Spatulas
Ecologically sustainable labware constructed of renewable raw materials
  • Packaged individually and suitable for food-grade applications
  • Produced and packaged according to ISO Class 7
  • EU foodstuffs and FDA compliant
The disposable sampling spatulas are long and V-shaped with a stable handle and sharp blade. This makes it possible to penetrate directly into containers such as paper or plastic sacks. These spatulas are ideal for sampling powders, granulates, and pastes. The total length of these spatulas is 26.3 cm with an insertion depth of 15 cm. These are available nonsterilized or presterilized via gamma irradiation.
$46.71 - $48.92CAD / Pkg of 10
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Overall Length (in) Sterile Description
Availability Pricing
Burkle 5379-0009 Disposable sampling spatula, PE, FDA compliant, white; 150 mm RK-06042-86 Mfr # 5379-0009
COMPARE
10 2/5 No Disposable sampling spatula, PE, FDA compliant, white; 150 mm
$46.71CAD / Pkg of 10
Burkle 5379-1009 Disposable sampling spatula, PE, FDA compliant, white, sterile; 150 mm RK-06042-93 Mfr # 5379-1009
COMPARE
10 2/5 Yes Disposable sampling spatula, PE, FDA compliant, white, sterile; 150 mm
$48.92CAD / Pkg of 10