This product is now discontinued, View Related Products
Bürkle Aluminum Spiralus Samplers for Powders and Granulates
Reliably sample various bulk goods, powders, and granulates for different sampling tasks
  • Efficient sampling made possible with the smallest gap possible between the inner and outer tube
  • Constructed of ultrapure materials for a durable design
These maintenance-free samplers allow for collection from the finest powders to large grains. They are suitable for sampling from all layers of the container, providing an accurate representation of the contents. These samplers come in different lengths, with single or multiple chambers and openings, and made of different material options for all different sampling methods. The 55 cm long samplers are suitable for sacks; the 85 cm long samplers are ideal for barrels, tubs, and mixed containers; and the 150 cm long samplers are appropriate for big bags, silos, and tanks.
Aluminum spiral samplers provide accurate representation of samples at each layer within the container. These are ideal for precise zone sampling as the spiral arrangement of the 8 openings open one after the other, from the lowest opening to the highest. The bottom of the sampler is filled first and then those above it in sequence are filled until the entire sampler is filled. These samplers are made of anodized aluminum with a PA tip, and feature one chamber with 8 openings.
$1,145.19 - $1,537.48CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Length (in) Volume (mL) Description
Availability Pricing
Burkle 5318-0145 Zone Sampling Lance, Aluminum/PA, 8 openings, one chamber; 770 mL capacity RK-06041-54 Mfr # 5318-0145
COMPARE
57 770 Zone sampling lance, aluminum/PA, 8 openings with one chamber, 145 cm length; 770 mL capacity
$1,145.19CAD / Each
Burkle 5318-0200 Zone Sampling Lance, Aluminum/PA, 8 openings, one chamber; 1100 mL capacity RK-06041-55 Mfr # 5318-0200
COMPARE
78.7 1100 Zone sampling lance, aluminum/PA, 8 openings with one chamber, 200 cm; 1100 mL capacity
$1,537.48CAD / Each