This product is now discontinued, View Related Products
B&K Precision Programmable DC Power Supplies
Easy-to-use programmable models at an economical price
  • Sixteen user programmable preset outputs
  • Controllable output on/off switch
  • 10 mV / 10 mA setable resolution
  • Overvoltage, overcurrent, and overtemperature protection
  • Closed case calibration
  • Serial interface cable and software for remote control operation
  • Low ripple and noise
  • Precise temperature stability
These programmable DC power supplies provide expanded programmable functionality with an intuitive front-panel interface making them easy to set up and operate. Units allow you to quickly and accurately make voltage and current selections with sixteen user-programmable preset outputs. The in-case calibration allows for simple, uninterrupted operation at an economical price.
$751.43 - $1,000.43CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Output Current Output Voltage Ripple and Noise
Availability Pricing
B&K Precision 1785B Programmable DC Power Supply, 18V, 5A RK-20045-92 Mfr # 1785B
COMPARE
0 to 5 A 0 to 18 VDC ≤ 1 mVrms, ≤ 3 mVp-p
$759.53CAD / Each
B&K Precision 1786B Programmable DC Power Supply, 32V, 3A RK-20045-93 Mfr # 1786B
COMPARE
0 to 3 A 0 to 32 VDC ≤ 1 mVrms, ≤ 3 mVp-p
$759.53CAD / Each
B&K Precision 1787B Programmable DC Power Supply, 72V, 1.5A RK-20045-94 Mfr # 1787B
COMPARE
0 to 1.5 A 0 to 72 VDC ≤ 1 mVrms, ≤ 3 mVp-p
$751.43CAD / Each
B&K Precision 1788 Programmable DC Power Supply, 32V, 6A RK-20045-95 Mfr # 1788
COMPARE
0 to 6 A 0 to 32 VDC ≤ 1 mVrms, ≤ 3 mVp-p
$1,000.43CAD / Each