This product is now discontinued, View Related Products
B&K Precision Programmable DC Electronic Loads
characterize DC power supplies, DC/DC converters, batteries, fuel cells, and solar cells
  • Constant current (CC), resistance (CR), voltage (CV), and power (CP) operation
  • Built-in transient generator with short circuit test
  • Built-in high resolution (0.1 mA/1 mV) voltage and current measurement (model 8500)
  • Over-current, over-voltage, over-power, and over-temperature protection
  • Flexible triggering: Create trigger events by front panel keystroke, back panel TTL signal, or software
  • Remote voltage sensing to compensate for the effect of voltage drop in wires
  • Store up to 25 instrument setups
5 PRODUCT OPTIONS
Compareitem specifications
Output Current Output Voltage Ripple and Noise
Availability Pricing
B&K Precision 8500 DC Electronic Load, 300W, 120V, 30A RK-20033-65 Mfr # 8500
COMPARE
0.001 to 30 A 0 to 120 VDC
$1,745.97CAD / Each
B&K Precision 8510 DC Electronic Load, 600W, 120V, 120A RK-20033-66 Mfr # 8510
COMPARE
0 to 120 A 0 to 120 VDC
$3,359.33CAD / Each
B&K Precision 8520 DC Electronic Load, 2400W, 120V, 240A RK-20033-67 Mfr # 8520
COMPARE
0 to 240 A 0 to 120 VDC
$8,530.93CAD / Each
B&K Precision IT-E151 Rack Mount Kit Model for Electronic Loads RK-20033-68 Mfr # IT-E151
COMPARE
$119.34CAD / Each
B&K Precision IT-E132 TTL to USB Serial Adapter, Model RK-20033-69 Mfr # IT-E132B
COMPARE
$75.14CAD / Each