This product is now discontinued, View Related Products
B&K Precision 4060 Series Dual-Channel Function/Arbitrary Waveform Generators
Generate stable and precise sine, square, triangle, pulse and arbitrary waveforms
  • Large 4.3” (109 mm) color display allows you to easily read current selected channel and parameters
  • Adjustable duty cycle and internal/external triggering provide additional flexibility
  • Store and recall up to 10 settings and 32 user-defined waveforms for quick and easy operation
  • GPIB connectivity with optional USB-to-GPIB adapter
These function/arbitrary generators are designed for applications that require high signal fidelity, a variety of modulation schemes, or arbitrary waveform generation capabilities such as education, research and design, and production test. Units feature two independent channels, with one-button synchronization, that can generate sine wave up to 160 MHz and provide linear/logarithmic sweep, built-in counter, variable DC offset, and extensive modulation triggering capabilities.
Use the included waveform editing software to create custom waveforms or use any of the 36 built-in predefined arbitrary waveforms that are easy to set up and navigate via the intuitive color interface with numeric keypad. The standard external 10 MHz reference clock input and output allow simple synchronization of these units with other function generator—and the phase of both output channels can be synchronized with the push of a button.
$306.47 - $3,505.19CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Max Frequency Range (Hz) Number Of Channels Wave Form
Availability Pricing
B&K Precision 4063 Function Generator, 2 Channel, 80 MHz RK-20048-24 Mfr # 4063
COMPARE
2 Pulse, ramp, sine, square, triangle
$2,213.03CAD / Each
B&K Precision 4064 Function Generator, 2 Channel, 120 MHz RK-20048-25 Mfr # 4064
COMPARE
2 Pulse, ramp, sine, square, triangle
$2,954.15CAD / Each
B&K Precision 4065 Function Generator, 2 Channel, 160 MHz RK-20048-26 Mfr # 4065
COMPARE
2 Pulse, ramp, sine, square, triangle
$3,505.19CAD / Each
B&K Precision AK40G USB to GPIB Adapter Cable RK-20048-27 Mfr # AK40G
COMPARE
$306.47CAD / Each