Aven Tools Cyclops Digital Microscopes
View magnified images on a large screen for user comfort and added productivity
  • HDMI output allows easy connection to an HD monitor
  • 2 megapixel sensor offers exceptionally crisp, high-resolution images
  • 30 white LEDs provide bright illumination
  • Included 11" high aluminum stand for large working distance
Minimize user fatigue from hunching over microscope eyepieces by connecting the Cyclops digital inspection microscope to a monitor. A convenient HDMI output allows quick connection to an HD monitor to produce a sharp, high-resolution image. Examine objects of various size is made easy with 30 adjustable intensity LEDs and a large working distance. Control focus and lighting remotely using an infrared remote controller. To connect to a PC order Cyclops 3.0 with USB 3.0 Type C output.
$1,466.02CAD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Magnification Output
Availability Pricing
Aven Tools Cyclops Digital Microscope, HDMI, 2 MP, 12X to 132X; 100 to 240 VAC RK-48708-32 Mfr # 26700-401
COMPARE
12 to 132x HDMI
$1,466.02CAD / Each