This product is now discontinued, View Related Products
Atago In-line Brix Refractometer
Continuously measure concentration of liquid samples
  • Digital display eliminates reading errors
This inline refractometer is ideal for use in the control of mixing and concentrating in food plants, fermentation processes, concentration of detergents, and more! Automatic temperature compensation allows measurements of high-temperature inline samples while ensuring accuracy. Control processing equipment or download data with the 4-20 mA and RS-232 outputs. The IP64-rated housing is dust tight and splash resistant. The sapphire prism and 316 stainless steel stage are the only materials in contact with the solution.
$514.95 - $7,098.79CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Accuracy Refractive Index Interval Resolution
Availability Pricing
Atago In-line Digital Brix Refractometer, 0 to 78%; 24 VDC RK-81005-25 Mfr # CM-780N
COMPARE
±0.2% (Brix) 0.1% (Brix)
$7,098.79CAD / Each
Atago Sample Inlet Unit, 12-mm barbed connections RK-81005-31 Mfr # RE-6645
COMPARE
$514.95CAD / Each
Atago In-Line Refractometer AC Power Supply/Adapter; 120 VAC RK-81005-33 Mfr # 3528
COMPARE
$980.91CAD / Each
Atago In-Line Refractometer AC Power Supply/Adapter; 240 VAC RK-81005-34 Mfr # 3529
COMPARE
$977.96CAD / Each