Atago® Digital Handheld Pocket Refractometers
Stop estimating readings
  • Simple operation—apply sample, press start, and reading is displayed in seconds
  • Extended range for added flexibility
  • Rugged design is IP65-rated and easy to clean
  • Easy calibration—just clean prism, add water, press the zero key
Compact, durable unit provides simple and accurate Brix readings
Lightweight and compact pocket refractometers are ideal for determining Brix concentrations in fruits and beverages, cutting oils, and organic samples. Use in the field or indoors, measurements are quick and accurate.
$764.69CAD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Brix Range (%) Min Brix Range (%) Resolution
Availability Pricing
Atago 3810 (PAL-1) Digital Pocket Refractometer, 0-53% Brix RK-02941-51 Mfr # 3810 (PAL-1)
COMPARE
53 0 0.1% (Brix)
$764.69CAD / Each