Ashcroft® G2 Stainless Steel Pressure Transducers
Accurate and reliable performance in a compact, durable package
  • ±1% total error band accuracy
  • High EMI/RFI rating
  • High temperature capability
High accuracy is accomplished by marrying a high performance ASIC to a very stable polysilicon thin film pressure sensor. The sensor is electron-beam welded to a pressure fitting of stainless steel, providing high overpressure capability and outstanding durability against shock and vibration. Transducer circuitry is held securely within an internal cage and housed in a reinforced nylon enclosure.
14 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Ashcroft G2 Compound Pressure Transducer, 30" Hg to 30 psi, 4 to 20 mA RK-68900-48 Mfr # G2-7-M02-42-G2-30#&VAC-G
COMPARE
Compound Pressure Transducer, 30\" Hg to 30 psi, 4 to 20 mA
$293.20CAD / Each
Ashcroft G2 Pressure Transducer, 0 to 30 psi, 4 to 20 mA RK-68900-50 Mfr # G2-7-M02-42-G2-30#-G
COMPARE
Pressure Transducer, 0 to 30 psi, 4 to 20 mA
$293.20CAD / Each
Ashcroft G2 Pressure Transducer, 0 to 100 psi, 4 to 20 mA RK-68900-52 Mfr # G2-7-M02-42-G2-100#-G
COMPARE
Pressure Transducer, 0 to 100 psi, 4 to 20 mA
$293.20CAD / Each
Ashcroft G2 Pressure Transducer, 0 to 200 psi, 4 to 20 mA RK-68900-54 Mfr # G2-7-M02-42-G2-200#-G
COMPARE
Pressure Transducer, 0 to 200 psi, 4 to 20 mA
$293.20CAD / Each
Ashcroft G2 Pressure Transducer, 0 to 500 psi, 4 to 20 mA RK-68900-56 Mfr # G2-7-M02-42-G2-500#-G
COMPARE
Pressure Transducer, 0 to 500 psi, 4 to 20 mA
$293.20CAD / Each
Ashcroft G2 Pressure Transducer, 0 to 1000 psi, 4 to 20 mA RK-68900-58 Mfr # G2-7-M02-42-G2-1000#-G
COMPARE
Pressure Transducer, 0 to 1000 psi, 4 to 20 mA
$293.20CAD / Each
Ashcroft G2 Pressure Transducer, 0 to 3000 psi, 4 to 20 mA RK-68900-60 Mfr # G2-7-M02-42-G2-3000#-G
COMPARE
Pressure Transducer, 0 to 3000 psi, 4 to 20 mA
$293.20CAD / Each
Ashcroft G2 Compound Pressure Transducer, 30" Hg to 30 psi, 1 to 5 VDC RK-68900-62 Mfr # G2-7-M02-15-G2-30#&VAC-G
COMPARE
Compound Pressure Transducer, 30\" Hg to 30 psi, 1 to 5 VDC
$293.20CAD / Each
Ashcroft G2 Pressure Transducer, 0 to 30 psi, 1 to 5 VDC RK-68900-64 Mfr # G2-7-M02-15-G2-30#-G
COMPARE
Pressure Transducer, 0 to 30 psi, 1 to 5 VDC
$293.20CAD / Each
Ashcroft G2 Pressure Transducer, 0 to 100 psi, 1 to 5 VDC RK-68900-66 Mfr # G2-7-M02-15-G2-100#-G
COMPARE
Pressure Transducer, 0 to 100 psi, 1 to 5 VDC
$293.20CAD / Each
Ashcroft G2 Pressure Transducer, 0 to 200 psi, 1 to 5 VDC RK-68900-68 Mfr # G2-7-M02-15-G2-200#-G
COMPARE
Pressure Transducer, 0 to 200 psi, 1 to 5 VDC
$293.20CAD / Each
Ashcroft G2 Pressure Transducer, 0 to 500 psi, 1 to 5 VDC RK-68900-70 Mfr # G2-7-M02-15-G2-500#-G
COMPARE
Pressure Transducer, 0 to 500 psi, 1 to 5 VDC
$293.20CAD / Each
Ashcroft G2 Pressure Transducer, 0 to 1000 psi, 1 to 5 VDC RK-68900-72 Mfr # G2-7-M02-15-G2-1000#-G
COMPARE
Pressure Transducer, 0 to 1000 psi, 1 to 5 VDC
$293.20CAD / Each
Ashcroft G2 Pressure Transducer, 0 to 3000 psi, 1 to 5 VDC RK-68900-74 Mfr # G2-7-M02-15-G2-3000#-G
COMPARE
Pressure Transducer, 0 to 3000 psi, 1 to 5 VDC
$293.20CAD / Each
Showing 1- 14 of 14