Ashcroft® Duragauge Plus! Process Pressure Gauges
Exclusive dry gauge gives liquid-filled performance and ±0.5% accuracy!
  • Revolutionary design reduces the effects of vibration and pulsation
  • No more liquid-fill lines—pointers are steady and readable
  • Eliminates the need for specialty fills—provides real cost savings
Gauges feature and 4 1/2" phenolic case for superior chemical and burn resistance, and a solid front case with a blow-out back for safety. Dry case gauge ensures no leakage, disposal, or environmental issues with any liquid fill. The case ring on all gauges is threaded polycarbonate. The back cover for all dry case gauges is polypropylene and for liquid filled and hermetic cases the cover is polycarbonate. Gauge is easily recalibrated.
$427.28 - $772.06CAD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Accuracy Max Pressure (PSI) Resolution
Availability Pricing
Ashcroft 1279SS4.5 Process Gauge 30" Hg to 15 psi RK-68039-00 Mfr # 45-1279-SS-04L-XLL-30IHGV/15#
COMPARE
±0.5% full-scale (ANSI/ASME B40.1 Grade 2A) 15 0.2\" Hg
$440.54CAD / Each
Ashcroft 1279SS4.5 Process Gauge 30" Hg to 30 psi RK-68039-02 Mfr # 45-1279-SS-04L-XLL-30IHGV/30#
COMPARE
±0.5% full-scale (ANSI/ASME B40.1 Grade 2A) 30 1\" Hg, 0.5 psi
$440.54CAD / Each
Ashcroft 1279SS4.5 Process Gauge 0 to 100 psi RK-68039-10 Mfr # 45-1279-SS-04L-XLL-100#
COMPARE
±0.5% full-scale (ANSI/ASME B40.1 Grade 2A) 100 1 psi
$427.28CAD / Each
Ashcroft 1279SS4.5 Process Gauge 0 to 200 psi RK-68039-12 Mfr # 45-1279-SS-04L-XLL-200#
COMPARE
±0.5% full-scale (ANSI/ASME B40.1 Grade 2A) 200 1 psi
$427.28CAD / Each
Ashcroft 1279SS4.5 Process Gauge 0 to 400 psi RK-68039-14 Mfr # 45-1279-SS-04L-XLL-400#
COMPARE
±0.5% full-scale (ANSI/ASME B40.1 Grade 2A) 400 1 psi
$427.28CAD / Each
Ashcroft 1279SS4.5 Process Gauge 0 to 600 psi RK-68039-16 Mfr # 45-1279-SS-04L-XLL-600#
COMPARE
±0.5% full-scale (ANSI/ASME B40.1 Grade 2A) 600 1 psi
$427.28CAD / Each
Ashcroft 1279SS4.5 Process Gauge 0 to 3000 psi RK-68039-20 Mfr # 45-1279SS-04L-XLL-3000#
COMPARE
±0.5% full-scale (ANSI/ASME B40.1 Grade 2A) 3000 20 psi
$444.96CAD / Each
Ashcroft 1279SS4.5 Process Gauge 0 to 20,000 psi RK-68039-26 Mfr # 45-1279-SS-04L-XLL-20000#
COMPARE
±0.5% full-scale (ANSI/ASME B40.1 Grade 2A) 20000 200 psi
$772.06CAD / Each
Showing 1- 8 of 8