Ashcroft® Digital Industrial Pressure Gauges
Three seperate styles of gauges - battery, loop-powered 4 to 20 mA, or line-powered 24 VDC
  • In-line powered unit acts as gauge, transducer, and switch
  • Intrinsically- safe IP65 case
  • Easy to read five digit display
Digital industrial gauge is perfect for pharmaceutical and biotech with its .25% terminal point accuracy. The multi-functionality of the gauge is great for chemical, pulp and paper, wastewater, and filtration processes, using local indication with the 4 to 20 mA output and switching options. Switch is rated for 125 VAC, 0.5 amp. Because the unit is digital, the switching option has unlimited deadband throughout the range of the gauge, and gives the unit superior set point flexability. Loop and in-line gauges have output signals that enable monitoring and control from remote areas such as control rooms or enclosures. Gauge range is vacuum to 20,000 psi.
Quarter percent accuracy includes linearity, hysteresis, repeatability, and termin
$980.54 - $1,530.85CAD / Each
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Ashcroft 4.5" Digital Pressure Gauge, Vacuum to 15 psi, Battery Powered RK-68338-00 Mfr # 45-2074-SD-02L-VAC/15#
COMPARE
4.5\" Digital Pressure Gauge, Vacuum to 15 psi, Battery Powered
$980.54CAD / Each
Ashcroft 4.5" Digital Pressure Gauge, Vacuum to 30 psi, Battery Powered RK-68338-02 Mfr # 45-2074-SD-02L-VAC/30#
COMPARE
4.5\" Digital Pressure Gauge, Vacuum to 30 psi, Battery Powered
$980.54CAD / Each
Ashcroft 4.5" Digital Pressure Gauge, Vacuum to 60 psi, Battery Powered RK-68338-04 Mfr # 45-2074-SD-02L-VAC/60#
COMPARE
4.5\" Digital Pressure Gauge, Vacuum to 60 psi, Battery Powered
$980.54CAD / Each
Ashcroft 4.5" Digital Pressure Gauge, 0 to 30 psi, Battery Powered RK-68338-06 Mfr # 45-2074-SD-02L-30#
COMPARE
4.5\" Digital Pressure Gauge, 0 to 30 psi, Battery Powered
$980.54CAD / Each
Ashcroft 4.5" Digital Pressure Gauge, 0 to 100 psi, Battery Powered RK-68338-10 Mfr # 45-2074-SD-02L-100#
COMPARE
4.5\" Digital Pressure Gauge, 0 to 100 psi, Battery Powered
$980.54CAD / Each
Ashcroft 4.5" Digital Pressure Gauge, 0 to 200 psi, Battery Powered RK-68338-12 Mfr # 45-2074-SD-02L-200#
COMPARE
4.5\" Digital Pressure Gauge, 0 to 200 psi, Battery Powered
$980.54CAD / Each
Ashcroft 4.5" Digital Pressure Gauge, 0 to 600 psi, Battery Powered RK-68338-14 Mfr # 45-2074-SD-02L-600#
COMPARE
4.5\" Digital Pressure Gauge, 0 to 600 psi, Battery Powered
$980.54CAD / Each
Ashcroft 4.5" Digital Pressure Gauge, 0 to 1000 psi, Battery Powered RK-68338-16 Mfr # 45-2074-SD-02L-1000#
COMPARE
4.5\" Digital Pressure Gauge, 0 to 1000 psi, Battery Powered
$980.54CAD / Each
Ashcroft 4.5" Digital Pressure Gauge, 0 to 10,000 psi, Battery Powered RK-68338-22 Mfr # 45-2074-SD-02L-10000#
COMPARE
4.5\" Digital Pressure Gauge, 0 to 10,000 psi, Battery Powered
$980.54CAD / Each
Ashcroft 4.5" Digital Pressure Gauge, Vacuum to 15 psi, Loop Powered RK-68338-28 Mfr # 45-2174-SD-02L-VAC/15#
COMPARE
4.5\" Digital Pressure Gauge, Vacuum to 15 psi, Loop Powered
$1,115.36CAD / Each
Ashcroft 4.5" Digital Pressure Gauge, Vacuum to 60 psi, Loop Powered RK-68338-32 Mfr # 45-2174-SD-02L-VAC/60#
COMPARE
4.5\" Digital Pressure Gauge, Vacuum to 60 psi, Loop Powered
$1,115.36CAD / Each
Ashcroft 4.5" Digital Pressure Gauge, 0 to 10,000 psi, Line Powered RK-68338-78 Mfr # 45-2274-SD-02L-XAO-XU1-XEN-10000#
COMPARE
4.5\" Digital Pressure Gauge, 0 to 10,000 psi, Line Powered
$1,530.85CAD / Each
Ashcroft 4.5" Digital Pressure Gauge, 30 to 0" Hg, Battery Powered RK-68338-84 Mfr # 45-2074-SD-02L-30IHGV
COMPARE
4.5\" Digital Pressure Gauge, 30 to 0\" Hg, Battery Powered
$980.54CAD / Each
Ashcroft 4.5" Digital Pressure Gauge, Vacuum to 100 psi, Battery Powered RK-68338-86 Mfr # 45-2074-SD-02L-VAC/100#
COMPARE
4.5\" Digital Pressure Gauge, Vacuum to 100 psi, Battery Powered
$980.54CAD / Each
Showing 1- 14 of 14