This product is now discontinued, View Related Products

Ashcroft Deadweight Tester kits

Primary pressure standard accurate to ±0.1% of reading
These self-contained tester kits have everything you need for testing, setting, calibrating, and repairing your high-accuracy pressure instruments—except operating fluid. Each kit consists of a two-stage hydraulic pump, two interchangeable piston assemblies (high- and low-pressure), a manifold with connectors, and an assortment of nonmagnetic, die cast zinc alloy weights. Precision-tooled weights are premeasured, marked with pressure values for each piston and come with a separate case.
Use the high-pressure piston for calibration pressures at maximum psi with a 75 psi resolution (see table at below); use the low-pressure piston to achieve 1/5 of the maximum pressure with a 15 psi resolution.
During operation, the manifold is pressurized until the weight assembly reaches a designated mark. A displacement valve lets you vary the pressure without using the pump handle or release valve. Once the correct pressure is achieved, simply reposition the needle on your gauge to the corresponding value.
$8,444.00 - $11,480.65CAD / Each

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Max Pressure Pressure Accuracy Pressure Resolution
Availability Pricing
Ashcroft 1305D Deadweight Tester Kit, 1000 psi Maximum RK-68840-00 Mfr # 1305D-10
COMPARE
1000 psi
$8,444.00CAD / Each
Ashcroft 1305D Deadweight Tester Kit, 2000 psi Maximum RK-68840-10 Mfr # 1305D-20
COMPARE
2000 psi
$8,837.39CAD / Each
Ashcroft 1305D Deadweight Tester Kit, 3000 psi Maximum RK-68840-20 Mfr # 1305D-30
COMPARE
3000 psi
$9,067.24CAD / Each
Ashcroft 1305D Deadweight Tester Kit, 5000 psi Maximum RK-68840-30 Mfr # 1305D-50
COMPARE
5000 psi
$9,363.39CAD / Each
Ashcroft 1305D Deadweight Tester Kit, 10,000 psi Maximum RK-68840-40 Mfr # 1305D-100
COMPARE
10000 psi
$11,480.65CAD / Each
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat