This product is now discontinued, View Related Products
Articulating Tip Fiberscope
Put visual control of hard-to-reach areas in your hands
  • Manueverable tip allows total viewing of inaccessible areas
These fiberscopes feature a control knob that allows the viewing tip to flex in directions needed for inspection. Internal fiber optic bundles provide sharp, high-resolution images of limited access areas. The small diameter shaft allows for inspection of internal components — eliminating or reducing equipment tear-down. Fiberscopes feature a shaft that keeps shape when bent or positioned during inspection.
$2,607.90 - $5,856.73CAD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Arm Length (in) Viewing Area Description
Availability Pricing
COMPARE
36 Articulating tip fiberscope; 36\"L x 7.0 mm OD
$4,059.19CAD / Each
COMPARE
24 Fiberscope, Two-way articulation, 24\" (609 mm)L, 0.276\" (7.0 mm) dia, +/-120° articulation
$3,631.91CAD / Each
COMPARE
24 Two-Way Articulating Tip Fiberscope, 24\"L, 0.236\" Dia, 120 Degree
$2,607.90CAD / Each
COMPARE
40 120 degrees Two-Way Articulating Tip Fiberscope, 40\"L, 0.236\" Dia, 120 Degree
$5,495.74CAD / Each
COMPARE
24 Fiberscope, Two-way articulation, 24\" (609 mm)L, 0.140\" (3.6 mm) dia, +/-100° articulation
$5,414.71CAD / Each
COMPARE
36 Fiberscope, Two-way articulation, 36\" (914 mm)L, 0.140\" (3.6 mm)dia, +/-100° articulation
$5,856.73CAD / Each
COMPARE
24 Four-Way Articulating Tip Fiberscope, 24\"L, 0.236\" Dia, 180 Degree
$2,755.24CAD / Each
COMPARE
40 180 degrees Four-Way Articulating Tip Fiberscope, 40\"L, 0.236\" Dia, 180 Degree
$5,206.96CAD / Each
Showing 1- 8 of 8