Cole-Parmer® Essentials Reusable Inoculating Streakers

Nichrome wire composition cools quickly and is less expensive than platinum
  • Select, inoculate, or transfer microbiological samples
Loops are made of high quality twisted nichrome wire and attached to a light-weight aluminum handle. Handles are insulated to protect the hand during use. Loops have been designed to slide along agar surfaces without cutting or tearing. Loops and Handles cool rapidly after sterilization and are offered calibrated (1µl & 10µl) or non-calibrated. Non-calibrated Loops and Handles are available as small, medium or large and are color coded for easy identification.
$103.14 - $374.24CAD / Pkg of 5

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Inoculating Streaker Loops, Small, Blue; 2.5 mm ID; 5/Pk
$103.14CAD / Pkg of 5
COMPARE
Inoculating Streaker Loops, Medium, Green; 3.5 mm ID; 5/Pk
$103.14CAD / Pkg of 5
COMPARE
Inoculating Streaker Loops, Large, Red; 5.5 mm ID; 5/Pk
$103.14CAD / Pkg of 5
COMPARE
Calibrated Inoculating Streaker Loops, 1ul, 1.45 mm ID; 5/PK
$374.24CAD / Pkg of 5
COMPARE
Calibrated Inoculating Streaker Loops, 10 ul, 4 mm ID; 5/PK
$374.24CAD / Pkg of 5
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat