Argos Technologies® PolarSafe® 100-Place EPS Microtube Racks
Secure, stackable freezer storage
  • Accommodates microtubes and cryovials up to 2.0 mL
  • Snug-fitting lid remains in place during handling
  • Grippable indentations allow easy removal from freezer
  • Alphanumeric index in base helps locate tubes
  • Store racks safely with the stacking system
This tube rack and storage box of expanded polystyrene (EPS) holds 100 microcentrifuge tubes and cryovials up to 2.0 mL. The deep wells and conical bottom keep tubes fixed in place, and the lid fits tightly enough to ensure tube safety during handling. Stacking system includes circular feet on the bottoms that mate with circular depressions on the lids. Centered indentations in the lid and base make removing a rack from the freezer easy, even if the rack is in the middle or bottom of the stack. Convenient alphanumerics located in the base keeps track of tubes.
$169.44CAD / Pkg of 10
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubes Held Tube Size Description
Availability Pricing
COMPARE
100 0.5 to 2.0 mL 100-Place EPS Microtube Rack; 10/pk
$169.44CAD / Pkg of 10