Argos Technologies® LiteSafe Microcentrifuge tubes
Protect photosensitive samples
  • Unlike amber tinted tubes, these are completely opaque protecting even the most sensitive samples
  • RNase and DNase free
Black tubes provide a completely opaque barrier, protecting even the most light sensitive compounds. Ideal for use with fluorescent probes or ethidium bromide. Tubes are made of biocompatible polypropylene with a click-fit cap to prevent accidental opening. Tubes can be stored down to -100°C and can be autoclaved to 122°C.
$43.47 - $45.68CAD / Pkg of 500
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tube Bottom Shape Tube Capacity (mL) Tube Material
Availability Pricing
COMPARE
Conical 0.5 Polypropylene
$43.47CAD / Pkg of 500
COMPARE
Conical 1.5 Polypropylene
$45.68CAD / Pkg of 500