Argos Technologies® Flip Cap Centrifuge Tubes
No more lost caps!
  • Hinged flip-top cap allows for single-handed use
  • Grooved cap surface for easier manipulation
  • Guaranteed DNase, RNase, and pyrogen free
  • Can be spun up to 9400 g
Centrifuge tubes with hinged flip-top caps allow for easy one-handed opening and closing. No more dropping or losing the tube cap. Bold black graduations are easy to read and accurate to ±2%. A large frosted marking area allows for permanent sample identification.
The conical-bottom, polypropylene (PP) tubes are compatible with any fixed angle or swing bucket rotor and can withstand temperatures from -80 to 120°C.
$198.91 - $262.26CAD / Case of 500
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Flip-Cap Centrifuge Tubes, 15 mL; 25/Bag, 20 Bags/Cs
$198.91CAD / Case of 500
COMPARE
Flip-Cap Centrifuge Tubes, 50 mL; 25/Bag, 20 Bags/Cs
$262.26CAD / Case of 500