Argos Technologies® ClickFit Microcentrifuge Tubes
Designed to prevent unintentional opening of lid during incubation or storage
  • Free of RNase, DNase, endotoxins, and human DNA
  • Certified TSE/BSE free
  • Manufactured in ISO class 8 cleanroom
  • Withstands centrifugation up to 30,000 g
ClickFit microcentrifuge tubes feature an increased opening force of the lid to prevent accidental cap opening keeping your samples safe and secure. The 1.5-mL polypropylene (PP) tube is autoclavable at 250F (121C) and freezable to -310F (-190C). Other features include an extended lip for easy opening, marked graduations (0.1, 0.5, 1.0, 1.5 mL), and frosted areas on tube and cap.
$27.26 - $46.41CAD / Case of 1000
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Cap Style Packaging Tube Capacity (mL)
Availability Pricing
COMPARE
$46.41CAD / Case of 1000
COMPARE
ClickFit 500/pk, 20,000/CS 6
$27.26CAD / Case of 500
COMPARE
ClickFit 500/pk, 8,000/CS 2
$44.57CAD / Case of 500