Argos Technologies® CapLock™ Microcentrifuge Tubes
Ideal for applications where sample integrity is importance
  • Free of RNase, DNase, endotoxins, and human DNA
  • Certified TSE/BSE free
  • Withstands centrifugation up to 30,000 g
CapLock™ microcentrifuge tubes features a double-lock cap closing system to ensure samples are safe and secure. It is ideal for boiling applications and working with DNA or hazardous substances. The polypropylene (PP) tube is autoclavable at 250°F (121°C) and freezable to -148°F (-100°C). Other features include an extended lip for easy opening, marked graduations, and frosted areas on tube and cap.
$33.52 - $58.94CAD / Pkg of 1000
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tube Capacity (mL) Description
Availability Pricing
COMPARE
1.5 CapLock Microcentrifuge Tube, 1.5 mL; 1000/Pk
$58.94CAD / Pkg of 1000
COMPARE
0.5 CapLock Microcentrifuge Tube, 0.5 mL; 500/Pk
$33.52CAD / Pkg of 500
COMPARE
2 CapLock Microcentrifuge Tubes, 2.0 mL, 500/PK
$47.52CAD / Pkg of 500