Applikon Fermentation Sampling System
Sampling system easily lets you remove a 60-mL sample from your fermentation vessel without removing the headplate. Attach system to the sample pipe via 1/4" ID tubing. Includes a 60-mL glass bottle with stainless steel fittings; optional 30-mL bottle is available, as well as additional 60-mL bottles.
$83.98 - $1,433.61CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Material Description
Availability Pricing
Applikon Fermentation Sampling System with 60 mL Bottle RK-01919-19 Mfr # Z81207SS02
COMPARE
Glass and Stainless Steel Fermentation Sampling System with 60 mL Bottle
$1,433.61CAD / Each
Applikon Replacement Glass Bottle for Fermenation Sampling System; 60 mL RK-01919-44 Mfr # Z81207BT60
COMPARE
Replacement Glass Bottle for Fermenation Sampling System; 60 mL
$83.98CAD / Each