This product is now discontinued, View Related Products
Ansell Vinyl Coat Apron
Also protects shoulders and arms
These coat aprons are made of FDA compliant materials — ideal for use in food handling applications. Seamless design eliminates contaminant traps while permitting cleaning to Class 100 standards. Coat aprons feature an open back/sleeves, no elastic, and have 40-mil PVC grommets permanently welded to the apron for greater strength and durability.
$20.63 - $46.04CAD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Color Length (in) Size
Availability Pricing
Ansell 950161 56-901 Vinyl Coat Apron; Thickness, 6-mil; Size, Large RK-81620-50 Mfr # 950161 56-901
COMPARE
Blue 57 Large
$20.63CAD / Each
Ansell 950162 56-901 Vinyl Coat Apron; Thickness, 6-mil; Size, X-large RK-81620-52 Mfr # 950162 56-901
COMPARE
Blue 57 X-Large
$20.63CAD / Each
Ansell 950191 56-910 Vinyl Coat Apron; Thickness, 8-mil; Size, Medium RK-81620-54 Mfr # 950191 56-910
COMPARE
Blue 57 Medium
$46.04CAD / Each
Ansell 950192 56-910 Vinyl Coat Apron; Thickness, 8-mil; Size, Large RK-81620-56 Mfr # 950192 56-910
COMPARE
Blue 57 Large
$46.04CAD / Each
Ansell 950193 56-910 Vinyl Coat Apron; Thickness, 8-mil; Size, X-large RK-81620-58 Mfr # 950193 56-910
COMPARE
Blue 57 X-Large
$46.04CAD / Each