This product is now discontinued, View Related Products
Amprobe Industrial True RMS Clamp Meters
Extra large jaw accommodates wide diameter wires
  • Peak Hold
  • 1000A DC current (model ACD-3400)
  • Relative zero and audible continuity
  • Auto and manual ranging with auto power off
  • Data hold
  • Backlight display
  • Diode test and duty cycle
  • Accommodates conductors up to 2 inches (51mm) in diameter
$456.74 - $1,035.78CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
DC Max (Volts) DC Voltage Accuracy True Rms
Availability Pricing
Amprobe AC68C DMM TRMS600 A AC/DC CLAMP-O RK-20003-29 Mfr # AC68C
COMPARE
$456.74CAD / Each
Amprobe ACD-3300 IND AC/DC Clamp Meter (1000A) RK-20034-08 Mfr # ACD-3300 IND
COMPARE
$742.57CAD / Each
Amprobe ACDC-3400 IND AC/DC Clamp Meter with DC Current RK-20034-09 Mfr # ACDC-3400 IND
COMPARE
$1,035.78CAD / Each