This product is now discontinued, View Related Products
Amprobe Heavy Duty Digital Multimeters
Built to be tough, reliable, safe and easy to use
  • Highest voltage capability, measuring to 1500 VDC and 1000 VAC
  • IP67 rated with special sealing for environment and water resistance
  • Constructed with drop-resistant housing and PCB mounting
  • CAT IV 1000 V rated
  • Auto power-down after 30 minutes of inactivity
  • Low battery indication
Designed for professional use in harsh environments, the heavy duty multimeters feature waterproof construction to prevent dirt, grime and moisture from entering the case and causing inaccurate readings or damaging shorts. Built of tough, fire-retardant thermoplastic, the multimeters are resistant to grease and chemical spills, with all sensitive components shock-mounted for a drop resistant to at least eight feet.
The extended voltage measurement capability of 1500 VDC and 1000 VAC is unmatched by any other line of digital multimeters. The extended voltage ranges can withstand transients up to 12 kV.
$76.60 - $861.92CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
DC Max (Volts) Resistance Max (Ω) Description
Availability Pricing
Amprobe HD160C Heavy Duty TRMS Digital Multimeter with Temperature RK-20005-77 Mfr # HD160C
COMPARE
1500 Heavy Duty TRMS Digital Multimeter with Temperature
$861.92CAD / Each
Amprobe DC205C Deluxe Hard Shell Carrying Case RK-20008-11 Mfr # DC205C
COMPARE
Deluxe Hard Shell Carrying Case
$76.60CAD / Each
Amprobe HD110C Heavy-Duty Digital Multimeter RK-23022-52 Mfr # HD110C
COMPARE
1500 Heavy-Duty Digital Multimeter
$528.93CAD / Each