This product is now discontinued, View Related Products

Air Tools - Sanders

  • Different pad shapes and sizes suitable for a wide range of applications
  • Ideal for sanding, polishing and buffing
Make the most of your air compressor with these air-powered tools. All require 90 psi pressure, but vary in air consumption.
$50.83 - $147.34CAD / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Free Air Capacity (CFM) Max Pressure (PSI) Description
Availability Pricing
Puma AT-7014 6 in Dual Action Sander, 5 mm orbital stroke, 6 in pad, 10,000 rpm, 1/4 in connection RK-07122-16 Mfr # AT-7014
COMPARE
16 90 6 in Dual Action Sander, 5 mm orbital stroke, 6 in pad, 10,000 rpm, 1/4 in connection
$88.40CAD / Each
Puma AT-7012 Air Tool, 5 in High Speed Sander, 5 in pad, 18,000 rpm, 1/4 in connection, 90 psi RK-07122-17 Mfr # AT-7012
COMPARE
4 90 Air Tool, 5 in High Speed Sander, 5 in pad, 18,000 rpm, 1/4 in connection, 90 psi
$50.83CAD / Each
Puma AT-7018 Air Orbital Sander, 3-2/3 in x 6-3/4 in pad, 8,000 rpm, 1/4 in connection, 90 psi RK-07122-18 Mfr # AT-7018
COMPARE
6 90 Air Orbital Sander, 3-2/3 in x 6-3/4 in pad, 8,000 rpm, 1/4 in connection, 90 psi
$81.04CAD / Each
Puma AT-7019 Air Tool, Straight Line Sander, 16 in x 2-1/2 in pad, 2,500 rpm, 1/4 in connection RK-07122-19 Mfr # AT-7019
COMPARE
10 90 Air Tool, Straight Line Sander, 16 in x 2-1/2 in pad, 2,500 rpm, 1/4 in connection
$147.34CAD / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat