This product is now discontinued, View Related Products
Air Tools - Impact / Ratchet Wrenches
  • Deliver high torque output with minimal exertion
  • Standard, butterfly, and pistol-grip styles available
Make the most of your air compressor with these air-powered tools. All require 90 psi pressure, but vary in air consumption and fittings. A variety of drive sizes, bolt capacities and maximum torque provide you with plenty of options.
$60.78 - $686.60CAD / Each
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Free Air Capacity (CFM) Max Pressure (PSI) Description
Availability Pricing
Puma AT-5030 Butterfly air impact wrench, 10,000 rpm, 75 ft-lb torque, 1/4 in connection, 90 psi RK-07122-01 Mfr # AT-5030
COMPARE
12 90 Butterfly air impact wrench, 10,000 rpm, 75 ft-lb torque, 1/4 in connection, 90 psi
$65.20CAD / Each
Puma AT-5033 Air impact wrench, 9,000 rpm, 75 ft-lb torque, 1/4 in connection, 90 psi RK-07122-02 Mfr # AT-5033
COMPARE
12 90 Air impact wrench, 9,000 rpm, 75 ft-lb torque, 1/4 in connection, 90 psi
$91.35CAD / Each
Puma AT-5133 Heavy Duty air impact wrench, 7,000 rpm, 200 ft-lb torque, 1/4 in connection, 90 psi RK-07122-03 Mfr # AT-5133
COMPARE
12 90 Heavy Duty air impact wrench, 7,000 rpm, 200 ft-lb torque, 1/4 in connection, 90 psi
$204.80CAD / Each
Puma AT-5040 Air impact wrench, 1/2 drive, 5/8 in bolt capacity, 7,000 rpm RK-07122-04 Mfr # AT-5040
COMPARE
20 90 Air impact wrench, 1/2 drive, 5/8 in bolt capacity, 7,000 rpm
$72.93CAD / Each
Puma AT-5340 Air impact wrench, 6,500 rpm, 400 ft-lb torque, 1/4 in connection, 90 psi RK-07122-06 Mfr # AT-5340
COMPARE
11.5 90 Air impact wrench, 6,500 rpm, 400 ft-lb torque, 1/4 in connection, 90 psi
$125.24CAD / Each
Puma AT-5342 Air impact wrench, 9,000 rpm, 550 ft-lb torque, 1/4 in connection, 90 psi RK-07122-07 Mfr # AT-5342
COMPARE
15 90 Air impact wrench, 9,000 rpm, 550 ft-lb torque, 1/4 in connection, 90 psi
$184.17CAD / Each
Puma AT-5060L Air impact wrench, 500 ft-lb torque, 1/4 in connection, 90 psi RK-07122-08 Mfr # AT-5060L
COMPARE
26 90 Air impact wrench, 500 ft-lb torque, 1/4 in connection, 90 psi
$201.85CAD / Each
Puma AT-5162 Air impact wrench, 5,000 rpm, 1,000 ft-lb torque, 3/8 in connection, 90 psi RK-07122-09 Mfr # AT-5162
COMPARE
19 90 Air impact wrench, 5,000 rpm, 1,000 ft-lb torque, 3/8 in connection, 90 psi
$315.31CAD / Each
Puma AT-5281L Air impact wrench 3,500 rpm, 1,600 ft-lb torque, 1/2 in connection, 90 psi RK-07122-11 Mfr # AT-5281L
COMPARE
36 90 Air impact wrench 3,500 rpm, 1,600 ft-lb torque, 1/2 in connection, 90 psi
$686.60CAD / Each
Puma AT-5051 Air impact wrench, 1/4 in drive, 1/4 in bolt capacity, 240 rpm, 15 ft-lb torque RK-07122-12 Mfr # AT-5051
COMPARE
6 90 Air impact wrench, 1/4 in drive, 1/4 in bolt capacity, 240 rpm, 15 ft-lb torque
$75.88CAD / Each
Puma AT-5052 Air impact wrench, 3/8 in drive, 3/8 in bolt capacity, 160 rpm, 50 ft-lb torque RK-07122-13 Mfr # AT-5052
COMPARE
8 90 Air impact wrench, 3/8 in drive, 3/8 in bolt capacity, 160 rpm, 50 ft-lb torque
$60.78CAD / Each
Puma AT-5054 Air impact wrench, 1/2 in drive, 1/2 in bolt capacity, 150 rpm RK-07122-14 Mfr # AT-5054
COMPARE
10 90 Air impact wrench, 1/2 in drive, 1/2 in bolt capacity, 150 rpm
$194.49CAD / Each
Showing 1- 12 of 12