Air/N2 Gas Mass Flowmeters with Metal Bodies
Precisely measure flow rates and totals
  • Proven design uses metal body components for added durability
  • Accumulated flow can be logged using an optional totalizer
  • Includes NIST-traceable certificate
These flowmeters feature an advanced U-shaped sensor that ensures accurate and repeatable
results. Gas flow measurements are unaffected by moderate temperature and pressure variations
at the inlet. The meter also allows a four-point calibration across the flow range to improve meter
linearity. Output data from the meter can be sent via a 0 to 5 VDC or 4 to 20 mA signal; an analogto-
RS converter is available for data collection and analysis on your PC.
All meters include a detachable 3 1/2-digit LCD that can be tilted up to 90° for easy reading.
Remotely mount the display up to three feet using extension cable 32662-70. The optional totalizer
32650-22 displays accumulated flow. The meter is protected from polarity reversal or short circuit
by a built-in resettable fuse. Aluminum models have wetted materials of anodized aluminum,
brass, and Viton®; stainless steel (SS) models have wetted materials of SS and Viton.
Accuracy:
These precalibrated flow controllers operate at inlet pressures between 5 and 60 psi and at gas temperatures between 59 and 77°F (15 to 25°C) while maintaining the stated percent full-scale accuracy and linearity. When operating beyond 5 to 60 psi, add ±0.01%/psi full-scale; if operating beyond 59 to 77°F (15 to 25°C), add ±0.15%/°C full-scale.
$957.70 - $2,927.63CAD / Each
31 variations of this product are available.
See All Product Options Below
31 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (LPM) Process Connection Wetted Materials
Availability Pricing
Aalborg GFMS-010001 Compact Gas Mass Flowmeter, 0-10 sccm, Air/N2, Aluminum Body RK-32648-00 Mfr # GFMS-010001
COMPARE
0.01 1/4" Compression Fittings Anodized Aluminum, Brass, Viton
$957.70CAD / Each
Aalborg GFMS-010002 Compact Gas Mass Flowmeter, 0-20 sccm, Air/N2, Aluminum Body RK-32648-02 Mfr # GFMS-010002
COMPARE
0.02 1/4" Compression Fittings Anodized Aluminum, Brass, Viton
$957.70CAD / Each
Aalborg GFMS-010003 Compact Gas Mass Flowmeter, 0-50 sccm, Air/N2, Aluminum Body RK-32648-04 Mfr # GFMS-010003
COMPARE
0.05 1/4" Compression Fittings Anodized Aluminum, Brass, Viton
$957.70CAD / Each
Aalborg GFMS-010007 Compact Gas Mass Flowmeter, 0-1 LPM, Air/N2, Aluminum Body RK-32648-12 Mfr # GFMS-010007
COMPARE
1 1/4" Compression Fittings Anodized Aluminum, Brass, Viton
$957.70CAD / Each
Aalborg GFMS-010008 Compact Gas Mass Flowmeter, 0-2 LPM, Air/N2, Aluminum Body RK-32648-14 Mfr # GFMS-010008
COMPARE
2 1/4" Compression Fittings Anodized Aluminum, Brass, Viton
$957.70CAD / Each
Aalborg GFMS-010009 Compact Gas Mass Flowmeter, 0-5 LPM, Air/N2, Aluminum Body RK-32648-16 Mfr # GFMS-010009
COMPARE
5 1/4" Compression Fittings Anodized Aluminum, Brass, Viton
$957.70CAD / Each
Aalborg GFMS-011159 Compact Gas Mass Flowmeter, 0-15 LPM, Air/N2, Aluminum Body RK-32648-19 Mfr # GFMS-011159
COMPARE
15 1/4" Compression Fittings Anodized Aluminum, Brass, Viton
$1,199.34CAD / Each
Aalborg GFMS-010014 Compact Gas Mass Flowmeter, 0-50 LPM, Air/N2, Aluminum Body RK-32648-36 Mfr # GFMS-010014
COMPARE
50 1/4" Compression Fittings Anodized Aluminum, Brass, Viton
$1,199.34CAD / Each
Aalborg GFMS-010015 Compact Gas Mass Flowmeter, 0-100 LPM, Air/N2, Aluminum Body RK-32648-42 Mfr # GFMS-010015
COMPARE
100 3/8" Compression Fittings Anodized Aluminum, Brass, Viton
$1,473.39CAD / Each
Aalborg GFMS-010017 Compact Gas Mass Flowmeter, 0-500 LPM, Air/N2, Aluminum Body RK-32648-44 Mfr # GFMS-010017
COMPARE
500 1/2" Compression Fittings Anodized Aluminum, Brass, Viton
$1,860.89CAD / Each
Aalborg GFMS-010019 Compact Gas Mass Flowmeter, 0-10 sccm, Air/N2, Stainless Steel Body RK-32648-50 Mfr # GFMS-010019
COMPARE
0.01 1/4" Compression Fittings 316 Stainless Steel, Viton
$1,449.82CAD / Each
Aalborg GFMS-010022 Compact Gas Mass Flowmeter, 0-100 sccm, Air/N2, Stainless Steel Body RK-32648-56 Mfr # GFMS-010022
COMPARE
0.1 1/4" Compression Fittings 316 Stainless Steel, Viton
$1,449.82CAD / Each
Aalborg GFMS-010023 Compact Gas Mass Flowmeter, 0-200 sccm, Air/N2, Stainless Steel Body RK-32648-58 Mfr # GFMS-010023
COMPARE
0.2 1/4" Compression Fittings 316 Stainless Steel, Viton
$1,449.82CAD / Each
Aalborg GFMS-010025 Compact Gas Mass Flowmeter, 0-1 LPM, Air/N2, Stainless Steel Body RK-32648-62 Mfr # GFMS-010025
COMPARE
1 1/4" Compression Fittings 316 Stainless Steel, Viton
$1,449.82CAD / Each
Aalborg GFMS-010027 Compact Gas Mass Flowmeter, 0-5 LPM, Air/N2, Stainless Steel Body RK-32648-66 Mfr # GFMS-010027
COMPARE
5 1/4" Compression Fittings 316 Stainless Steel, Viton
$1,449.82CAD / Each
Aalborg GFMS-011161 Compact Gas Mass Flowmeter, 0-15 LPM, Air/N2, Stainless Steel Body RK-32648-68 Mfr # GFMS-011161
COMPARE
15 1/4" Compression Fittings 316 Stainless Steel, Viton
$1,800.48CAD / Each
Aalborg GFMS-010030 Compact Gas Mass Flowmeter, 0-50 LPM, Air/N2, Stainless Steel Body RK-32648-86 Mfr # GFMS-010030
COMPARE
50 1/4" Compression Fittings 316 Stainless Steel, Viton
$1,800.48CAD / Each
Aalborg GFMS-010031 Compact Gas Mass Flowmeter, 0-100 LPM, Air/N2, Stainless Steel Body RK-32648-92 Mfr # GFMS-010031
COMPARE
100 3/8" Compression Fittings 316 Stainless Steel, Viton
$2,059.80CAD / Each
Aalborg GFMS-010803 Compact Gas Mass Flowmeter, 0-200 LPM, N2/Air, Stainless Steel Body RK-32659-00 Mfr # GFMS-010803
COMPARE
200 3/8" Compression Fittings 316 Stainless Steel, Viton
$2,439.93CAD / Each
Aalborg GFMS-010243 Compact Gas Mass Flowmeter, 0-500 LPM, N2/Air, Stainless Steel Body RK-32659-10 Mfr # GFMS-010243
COMPARE
500 1/2" Compression Fittings 316 Stainless Steel, Viton
$2,602.01CAD / Each
Showing 1 - 20 of 31