This product is now discontinued, View Related Products
AEMC Fall of Potential Ground Resistance Testers
FEATURES
  • Measures soil resistivity (4-point) (Model AE211494)
  • Measures ground resistance (2 and 3 point) fall of potential method
  • Large analog display (Model AE211490)
  • Large LCD digital display (Models AE211492 and AE211494)
  • Designed to reject high levels of noise and interference
  • Auto-ranging: automatically selects the optimum range (Models AE211492 and AE211494))
  • Battery powered
  • Extremely simple to operate: connect - press - read
  • Indicator error lights
  • Rugged dustproof and rainproof case
  • Color-coded terminals and lead identification
Model AE211495 Kit includes: meter, one 16 ft color-coded lead, two 100 ft color-coded leads, two 300 ft leads on reels, four 16" auxiliary ground electrodes, soft carrying case, batteries and user manual.
Model AE211493 Kit includes meter, one 16 ft lead, two 150 ft leads on reels with wind-up handles, two 16" auxiliary ground electrods, soft carrying case, batteries and user manual.
Model AE211494Kit includes meter, one 16 ft lead, two 150 ft leads on reels with wind-up handles, two 16" auxiliary ground electrods, soft carrying case, batteries and user manual.
$1,193.45 - $2,429.62CAD / Each
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
Compareitem specifications
AC Current Accuracy AC Current Range (mA) AC Current Resolution (mA)
Availability Pricing
AEMC 3620KIT-150FT Kit-150 ft Ground Resistance Tester RK-20014-94 Mfr # 3620KIT-150FT
COMPARE
10
$1,810.80CAD / Each
AEMC 3620KIT-300FT Kit-300 ft Ground Resistance Tester RK-20014-95 Mfr # 3620KIT-300FT
COMPARE
10
$1,958.14CAD / Each
AEMC 3620 Kit-500 ft Ground Resistance Tester RK-20014-96 Mfr # 3620KIT-500FT
COMPARE
10
$2,002.34CAD / Each
AEMC 3640KIT-150FT Kit-150 ft 150FT Ground Resistance Test Kit RK-20014-97 Mfr # 3640KIT-150FT
COMPARE
10
$1,812.27CAD / Each
AEMC 3640KIT-300FT Kit-300 ft Ground Resistance Tester RK-20014-98 Mfr # 3640KIT-300FT
COMPARE
10
$1,959.61CAD / Each
AEMC 3640 Kit-500 ft Ground Resistance Tester RK-20014-99 Mfr # 3640KIT-500FT
COMPARE
10
$2,025.91CAD / Each
AEMC 4610 Kit-150 ft Ground Resistance Tester RK-20015-00 Mfr # 4610KIT-150FT
COMPARE
10
$2,134.94CAD / Each
AEMC 4610 Kit-300 ft Ground Resistance Test Kit RK-20015-01 Mfr # 4610KIT-300FT
COMPARE
10
$2,282.28CAD / Each
AEMC 4610 Kit-500 ft Ground Resistance Tester RK-20015-02 Mfr # 4610KIT-500FT
COMPARE
10
$2,429.62CAD / Each
AEMC 3620 Analog Ground Resistance Ground Resistance Tester RK-20024-08 Mfr # 3620
COMPARE
10
$1,215.55CAD / Each
AEMC 3640 Digital Ground Resistance Digital Ground Resistance Tester RK-20024-09 Mfr # 3640
COMPARE
10
$1,193.45CAD / Each
AEMC 4610 Ground Resistance Tester Digital 4-Point- RK-20024-10 Mfr # 4610
COMPARE
$1,539.69CAD / Each
Showing 1- 12 of 12