This product is now discontinued, View Related Products
AEMC 6740B Series Multi-Function Ground Resistance Tester
Measure ground resistance, soil resistivity and bonding resistance with one instrument.
FEATURES
  • 3- and 4-Point measurement with manual or automatic frequency selection (even during measurement) from 40 up to 500Hz,>120dB noise reduction
  • Selectable test voltage of 16 or 32Vac RMS
  • True RMS (AC, DC, AC/DC) voltage measurement from 200mV to 50V
  • Automatic measurement of auxiliary electrodes resistance
  • Soil resistivity measurement using Wenner or Schlumberger test method
  • 2- and 4-Point DC resistance measurement with automatic polarity change (Bonding test function)
  • Auto-off for power management
  • Automatic recognition of all electrode connections and their resistance
  • Memory stores up to 512 test results
  • USB communication
  • Remote operation of all measurement functions using DataView® software
  • Automatic report generation
  • Rechargeable NiMH batteries
  • Rugged dustproof and rainproof field case — IP54 in closed position
  • IEC 61557 part 4 and part 5
  • Includes DataView® software for data storage, real-time display, analysis and report generation
Standard Accessories Include: meter, NiMH batteries, optically isolated USB cable, Dataview® software, external battery charger, AC power cord and user manual.
Multi-Function Ground Resistance Tester Model 6470 Kit includes: meter, NiMH batteries, optically isolated USB cable, DataView® software, external battery charger, AC power cord, one 30 ft green wire, one each 500 ft red and blue wire, one each 100 ft black and green wire, set of five fork terminals, four ground electrodes, tape measure, carrying case and user manual, Cat. No. AE213504
$3,506.67 - $4,582.24CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
AC Current Accuracy AC Current Range (mA) AC Current Resolution (mA)
Availability Pricing
AEMC 6470B B Ground Resistance Tester with Software RK-20014-90 Mfr # 6470-B
COMPARE
$3,506.67CAD / Each
AEMC 6470B-kit150 Ground Resistance Tester Kit with 150 Ft. Lead And Accessories RK-20014-91 Mfr # 6470KIT-150FT
COMPARE
$3,903.01CAD / Each
AEMC 6470B-kit300 Ground Resistance Tester Kit with 300 Ft. Lead And Accessories RK-20014-92 Mfr # 2135.03
COMPARE
$4,346.50CAD / Each
AEMC 6470B-kit500 Ground Resistance Tester Kit Tester RK-20014-93 Mfr # 2135.04
COMPARE
$4,582.24CAD / Each